missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 2 Antibody (SAIC-34D-11) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30167639
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30167639 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30167639 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP320103JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Peroxiredoxin 2 Monoclonal antibody specifically detects Peroxiredoxin 2 in Human samples. It is validated for Flow Cytometry,Western Blot,Mass Spectrometry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 2
Applications Flow Cytometry, Western Blot, Mass Spectrometry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone SAIC-34D-11
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1, MGC4104, Natural killer cell-enhancing factor B, natural killer-enhancing factor B, NKEFBNKEF-B, peroxiredoxin 2, peroxiredoxin-2, PRPTDPX1, PRX2, PRXII, thiol-specific antioxidant 1, Thiol-specific antioxidant protein, Thioredoxin peroxidase 1, Thioredoxin-dependent peroxide reductase 1, torin, TPX1, TSAEC 1.11.1.15
Host Species Rabbit
Immunogen Peptide 'DHPFGFVAVPTK', derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7001
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.