missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99736-50ul
This item is not returnable.
View return policy
Description
Peroxiredoxin 1 Polyclonal antibody specifically detects Peroxiredoxin 1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Peroxiredoxin 1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 uL/mg of lysate | |
| EC 1.11.1, EC 1.11.1.15, Natural killer cell-enhancing factor A, natural killer-enhancing factor A, NKEFA, NKEF-A, PAG, PAGAMSP23, PAGB, peroxiredoxin 1, peroxiredoxin-1, proliferation-associated gene A, Proliferation-associated gene protein, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2 | |
| Produced in rabbits immunized with purified, recombinant Human Peroxiredoxin 1 (Accession#: NP_002565.1; Met 1-Lys 199) | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5052 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction