missing translation for 'onlineSavingsMsg'
Learn More

Permethrinic Acid (cis/trans mixture), TRC

Product Code. 30009323
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009323 1 g 1g
30003404 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009323 Supplier Toronto Research Chemicals Supplier No. TRCD4343001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55701-05-8
Molecular Formula C8 H10 Cl2 O2
Molecular Weight (g/mol) 209.07
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, Cyclopropanecarboxylic acid, 3-(2,2-dichlorovinyl)-2,2-dimethyl- (6CI), 2,2-Dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylic acid, 3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropylcarboxylic acid, DV-chrysanthemic acid, Permethric acid, Permethrinic acid
IUPAC Name 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H10Cl2O2/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3,(H,11,12)
Formula Weight 208.0058
Percent Purity >95%
Chemical Name or Material 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic Acid (Permethrinic Acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.