missing translation for 'onlineSavingsMsg'
Learn More

Permethrin, MedChemExpress

Product Code. p-200096363
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16438486

Brand: Medchem Express HYB0887

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent; functions as a neurotoxin, affecting neuron membranes by prolonging sodium channel activation.

Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent

Specifications

Chemical Name or Material Permethrin
CAS 52645-53-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20Cl2O3
Quantity 100 mg
Synonym NRDC-143
Solubility Information DMSO : 50 mg/mL (127.78 mM; Need ultrasonic)
SMILES O=C(C1C(C)(C)C1/C=C(Cl)\Cl)OCC2=CC=CC(OC3=CC=CC=C3)=C2
Molecular Weight (g/mol) 391.29
Formula Weight 391.29
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.