missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Permethrin, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Permethrin |
| CAS | 52645-53-1 |
| CAS Max % | ≤ 100% |
| Density | 1.29 g/cm3 (20°C) |
| Flash Point | 100°C |
| Beilstein | 5765325 |
| Color | Beige |
| Melting Point | 34°C to 35°C |
| Boiling Point | 200°C (0.01 hPa) |
| Synonym | (3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropanecarboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction