missing translation for 'onlineSavingsMsg'
Learn More

Permethrin, Dr. Ehrenstorfer

Product Code. 30325749
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30325749

Brand: Dr. Ehrenstorfer DREC15990000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Permethrin
CAS 52645-53-1
Analyte or Component Names Permethrin
Shipping Condition +20°C
Molecular Formula C21 H20 Cl2 O3
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (3-phenoxyphenyl)methyl ester, (3-Phenoxyphenyl)methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, Acticin, Adion, Ambush, Aninsen Per-30, Anomethrin N, Antiborer 3768, Aqua-Kontrol, Astro, Bansect, Bematin 987, BioKill, Butox 50, Chinetrin, Cooper, Coopex, Corsair, Damminix, Dancide PS 150, Dichlorophenothrin, Diffusil H, Dragnet, Dragnet FT, Dragon, Ecsumin, Ectiban, Efmethrin, Eliminator Ant, Flea & Tick Killer, Elimite, Eulan SPA, Everwood PC 30W, Exmin, FMC 33297, FMC 41655, ICI-PP 557, Imperator, InfectoPedicul, Insectal Plus, Insorbcid MP, Ipitox, JF 7065, Kaleait, Kavil, Kestrel, Kestrel (pesticide), Kudos, Last Call, Lyclear, MP 79, MP 79 (ester), Mitin BC, NIA 33297, NRDC 143, Nedax, Nix, Nix (insecticide), PP 557, Perigen, Perigen 500, Perigen Defense, Perigen W, Perm-UP, Permanone, Permanone 40, Permanone 80, Permanone 90, Permasect, Permectin II, Permethrin, Permethrin I, Permethrin II, Permethrine, Permitrine, Pertrin E, Piredam, Pounce, Preventol HS 75, Pulvex, Pynosect, Repel Permanone, Riapan A, Ridect Pour-On, S 3151, SBP 1513, SBP 15131TEC, Sharkesuper, Skintex MR-III, Smite EC 10, Spartan, Spartan (pesticide), Stomoxin, Stomozan, Tanalith T, Timbertreat ICP 5, WL 43479, m-Methoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, m-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, GW418275X
InChI Formula InChI=1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2
IUPAC Name (3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 391.29
Quantity 100 mg
Formula Weight 390.0789 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.