Learn More
Invitrogen™ eBioscience™ Permeabilization Buffer (10X)
Product Code. 12766048
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
Product Code. Quantity Unit Size
12766048 100 mL Each
1 options
Contains Sodium azide
Product Code. 12766048 Supplier Invitrogen™ Supplier No. 00833356
  • ONLINE EXCLUSIVE PRICE
    £102.00
    £85.65 / 100mL
    Save £16.35
    16% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Contains Sodium azide
Can be used to fix and permeabilize cells prior to performing intracellular staining of cytokines and other cytoplasmic and/or nuclear antigens.
Used in conjunction with the Intracellular (IC) Fixation Buffer (cat. 00-8222) or Foxp3/Transcription Factor Fixation/Permeabilization Concentrate and Diluent (cat. 00-5521). These solutions have been specially formulated for reducing non-specific staining of fluorochrome-labeled antibodies and increasing fluorescence signal to noise ratios.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Reagent Type Permeabilization Reagent
Product Line eBioscience
Product Type Permeabilization Buffer