missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perlecan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5140142
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RGMVFGIPDG VLELVPQRGP CPDGHFYLEH SAACLPCFCF GITSVCQSTR RFRDQIRLRF DQPDDFKGVN VTMPAQPGTP PLSSTQLQID PSL
Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.Specifications
Perlecan | |
Polyclonal | |
Unconjugated | |
HSPG2 | |
AI852380; Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); heparan sulfate proteoglycan 2; HSPG; Hspg2; LG3 peptide; LOC313641; Pcn; per; perlecan; perlecan (heparan sulfate proteoglycan 2); perlecan proteoglycan; PLC; PRCAN; RP11-132G19.2; SJA; SJS; SJS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P98160 | |
HSPG2 | |
Recombinant protein corresponding to Human HSPG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |