missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Perlapine
GREENER_CHOICE

Product Code. 18795456
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18795456

Brand: Tocris Bioscience™ 5549/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent muscarinc DREADD agonist

Perlapine is a potent muscarinic DREADD agonist (pEC50 values are 8.38, 8.08 and 7.27 for hM1Dq, hM3Dq and hM4Di in vitro, respectively). Exhibits >10,000-fold selectivity for hM3Dq over hM3 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-(4-Methyl-1-piperazinyl)-11 H-dibenz[b,e]azepine
CAS 11-3-1977
Quantity 10 mg
Target DREADD Activators
Molecular Formula C19H21N3
Formula Weight 291.39
Purity 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.