Learn More
Trajan Scientific and Medical™ PerkinElmer Series 200 LC Autosampler Syringe
Accurate and reproducible injections.
Brand:  Trajan Scientific and Medical™ 008185
Code : 13
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12760710
  • £111.00 / Each
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for PerkinElmer instruments
Specifications
Specifications
PTFE (Plunger)
High-performance Liquid Chromatography
1 mL
Gas Tight