missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PERK Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£171.00 - £401.00
Specifications
| Antigen | PERK |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232012
|
Novus Biologicals
NBP3-33306-20ul |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227218
|
Novus Biologicals
NBP3-33306-100ul |
100 μL |
£401.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PERK Monoclonal antibody specifically detects PERK in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PERK | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 9451 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 580-720 of human PERK (NP_004827.4).,, Sequence:, IKNSGYISRYLTDFEPIQCLGRGGFGVVFEAKNKVDDCNYAIKRIRLPNRELAREKVMREVKALAKLEHPGIVRYFNAWLEAPPEKWQEKMDEIWLKDESTDWPLSSPSPMDAPSVKIRRMDPFATKEHIEIIAPSPQRSR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title