missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PERK Polyclonal antibody specifically detects PERK in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot
Specifications
Specifications
| Antigen | PERK |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:4000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:150, Immunohistochemistry-Paraffin, Dot Blot 1:4000 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 1000-1050 on human Eukaryotic translation initiation factor 2-alpha kinase 3 protein. |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?