Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Product Code. p-6084926 Shop All Thermo Scientific Products
Click to view available options
Type:
Peristaltic Pump Tubing
Diameter (Metric):
1.295 mm
Material:
Santoprene
Unit Size:
Each
Product Code. 15948486
Brand: Thermo Scientific™ 1320110
This item requires a price update prior to being ordered.
Request a Price for this item here.
Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing for the iCAP™ MX and iCAP™ Q/Qnova Series is available to suit your sample matrices and ICP-MS applications.
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
  • Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
  • Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
  • Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
  • Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
  • Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
  • Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Type Peristaltic Pump Tubing
For Use With (Equipment) iCAP MX and iCAP Q/Qnova Series
Material Santoprene
Diameter (Metric) 1.295 mm