Learn More
Invitrogen™ Periplakin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120250
Description
Positive test controls include: Mouse lung, Mouse heart. The target is usually found in the following locations: Cell junction, Cell membrane, Cytoplasm, Mitochondrion, Nucleus, cytoskeleton, desmosome. Immunogen sequence: MNSLFRKRNK GKYSPTVQTR SISNKELSEL IEQLQKNADQ VEKNIVDTEA KMQSDLARLQ EGRQPEHRDV TLQKVLDSEK LLYVLEADAA IAKHMKHPQG DMIAEDIRQL KERVTNLRGK HKQIYRLAVK.
The protein encoded by this gene is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
Specifications
| Periplakin | |
| Polyclonal | |
| Unconjugated | |
| Ppl | |
| 190 kDa paraneoplastic pemphigus antigen; 195 kDa cornified envelope precursor protein; 195kDa protein; AW553870; KIAA0568; MGC134872; Periplakin; Ppl | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19041, 302934, 5493 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.99 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60437, Q9R269 | |
| Ppl | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PPL (NP_002696.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.