missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periplakin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95228-0.02ml
This item is not returnable.
View return policy
Description
Periplakin Polyclonal antibody specifically detects Periplakin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Periplakin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| 190 kDa paraneoplastic pemphigus antigen, 195 kDa cornified envelope precursor protein, KIAA0568, MGC134872, periplakin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PPL (NP_002696.3). MNSLFRKRNKGKYSPTVQTRSISNKELSELIEQLQKNADQVEKNIVDTEAKMQSDLARLQEGRQPEHRDVTLQKVLDSEKLLYVLEADAAIAKHMKHPQGDMIAEDIRQLKERVTNLRGKHKQIYRLAVK | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| 5493 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction