missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Peripherin Monoclonal Antibody (8G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA316724
This item is not returnable.
View return policy
Description
Peripherin Monoclonal Antibody for Western Blot, IHC, ICC/IF
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
Specifications
| Peripherin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.035% sodium azide | |
| A6QQJ3, P15331, P21807, P41219 | |
| PRPH | |
| Recombinant rat Peripherin purified from E. coli. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 8G2 | |
| Unconjugated | |
| PRPH | |
| if3; nef4; Neurofilament 4; neurofilament 4 (57kD); neuronal intermediate filament IF3; Perf; Peripherin; peripherin 1; peripherin L homeolog; plasticin; PRPH; prph.L; PRPH1; XELAEV_18013332mg; XIF3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100152434, 19132, 24688, 510082, 5630 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction