missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (OTI1E10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73337CL1
This item is not returnable.
View return policy
Description
Peripherin Monoclonal antibody specifically detects Peripherin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Peripherin | |
| Monoclonal | |
| CoraFluor 1 | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Full length human recombinant protein of human PRPH (NP_006253) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 5630 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1E10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction