missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Peripherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38070-20ul
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Peripherin Polyclonal antibody specifically detects Peripherin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Tekniske data
| Peripherin | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human Peripherin (NP_006253.2).,, Sequence:, MSHHPSGLRAGFSSTSYRRTFGPPPSLSPGAFSYSSSSRFSSSRLLGSASPSSSVRLGSFRSPRAGAGALLRLPSERLDFSMAEALNQEFLATRSNEKQELQELNDRFANFIEKVRFLEQQNAALRGELSQARGQEPARADQLCQQELRELRRELELLGRERDRVQVERDGLAEDLAALKQRLEEETRKREDAEHNLVLFRKDVDDATLSRLELERKIESLMDEIEFLKKLHEEELRDLQVSVESQQVQQVEVEATVKPE | |
| 20 μL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 5630 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion