missing translation for 'onlineSavingsMsg'
Learn More

Peripherin 2/PRPH2 Antibody [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30762183 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762183 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762183 Supplier Novus Biologicals Supplier No. NBP346880AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peripherin 2/PRPH2 Polyclonal antibody specifically detects Peripherin 2/PRPH2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin 2/PRPH2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AOFMD, AVMD, CACD2peripherin 2, homolog of mouse, DS, peripherin 2 (retinal degeneration, slow), peripherin-2, PRPHperipherin, photoreceptor type, rd2, RDS, Retinal degeneration slow protein, retinal degeneration, slow, retinal peripherin, RP7, Tetraspanin-22, tspan-22, TSPAN22retinal degeneration, slow (retinitis pigmentosa 7)
Host Species Rabbit
Immunogen peripherin 2(retinal degeneration, slow). (Uniprot# P23942)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5961
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.