missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periostin/OSF-2 Antibody (POSTN/8523R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20615-20ug
This item is not returnable.
View return policy
Description
Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Periostin/OSF-2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1 | |
| Recombinant fragment (around aa193-326) of human Periostin/OSF-2 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cardiovascular Biology, Extracellular Matrix | |
| 10631 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| POSTN/8523R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction