missing translation for 'onlineSavingsMsg'
Learn More

Periostin/OSF-2 Antibody (613643R) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30733641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733641 Supplier Novus Biologicals Supplier No. FAB35484MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periostin/OSF-2
Applications ELISA
Classification Monoclonal
Clone 613643R
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Periostin/OSF-2, Met1-Gln836, Accession # Q15063
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 10631
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.