missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Periostin Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550891
This item is not returnable.
View return policy
Description
Periostin (PN), also designated osteoblast-specific factor 2 (OSF-2), is a disulfide linked protein originally isolated as a osteoblast-specific factor. Periostin is a secreted protein that binds heparin and functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins. In preosteoblasts, Periostin acts as a cell adhesion molecule and plays a role in osteoblast recruitment, spreading and attachment. Periostin is mainly detected in lower gastrointestinal tract, aorta, stomach, placenta, uterus and breast tissues but is up-regulated in epithelial ovarian tumors and overexpressed in breast cancer. Expression of Periostin is increased by bone morphogenetic protein (BMP2) and transforming growth factor beta 1(TGF beta 1). Periostin contains a typical signal sequence, followed by a cysteine-rich domain, a fourfold repeated domain, which shows homology with the insect protein fasciclin, and a C-terminal domain.
Specifications
| Periostin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q15063 | |
| Postn | |
| Recombinant Human POSTN protein, Pro97-Leu230 (Accession #Q15063). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 κ |
| Western Blot, Immunocytochemistry, Western Blot | |
| C2 | |
| Unconjugated | |
| Postn | |
| A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10631, 361945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction