missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, Honeywell™

Product Code. p-7072307 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15698050

Brand: Honeywell Research Chemicals P787525G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
SMILES O[I](O)(O)(O)(O)=O

Specifications

Quantity 25 g
UN Number UN3085
Formula Weight 227.94g/mol
Percent Purity ≥99%
Chemical Name or Material Periodic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.