missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, 99+%, extra pure

Product Code. p-4509555 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10592981

Brand: Thermo Scientific Acros 198401000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid | CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White to Yellow
Melting Point 122.0°C
Identification Positive for periodate
Packaging Glass bottle
Assay Percent Range 99% min. (Iodometry)
Quantity 100 g
Linear Formula H5IO6
Physical Form Crystals or Crystalline Powder
Fieser 01,815; 02,313; 03,220; 04,374; 05,508; 10,304; 13,238; 16,272
Merck Index 15, 7286
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol,slightly soluble in ether
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Formula Weight 227.94
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Periodic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Periodic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.