Learn More
Periodic acid, 34% w/w aq. soln., Thermo Scientific Chemicals
£40.90 - £518.00
Chemical Identifiers
CAS | 10450-60-9 |
---|---|
Molecular Formula | H5IO6 |
Molecular Weight (g/mol) | 227.94 |
MDL Number | MFCD00011344 |
InChI Key | TWLXDPFBEPBAQB-UHFFFAOYSA-N |
Synonym | orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst |
PubChem CID | 25289 |
ChEBI | CHEBI:29150 |
IUPAC Name | pentahydroxy(oxo)-$l^{7}-iodane |
SMILES | O[I](O)(O)(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479596
|
Thermo Scientific Alfa Aesar
B20172.22 |
100 g |
£40.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489596
|
Thermo Scientific Alfa Aesar
B20172.36 |
500 g |
£156.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499596
|
Thermo Scientific Alfa Aesar
B20172.0E |
2500 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10450-60-9 | |
227.94 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
25289 | |
pentahydroxy(oxo)-$l^{7}-iodane |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
CHEBI:29150 | |
O[I](O)(O)(O)(O)=O |
Specifications
10450-60-9 | |
H5IO6 | |
UN3098 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
O[I](O)(O)(O)(O)=O | |
227.94 | |
CHEBI:29150 | |
Periodic acid |
34% w/w aq. soln. | |
MFCD00011344 | |
14,7171 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
pentahydroxy(oxo)-$l^{7}-iodane | |
25289 | |
227.94 |
Safety and Handling
GHS H Statement
H272-H314
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only