missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perindopril t-Butylamine Salt, TRC
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
Specifications
Specifications
| Chemical Name or Material | Perindopril tert-Butylamine |
| CAS | 107133-36-8 |
| Molecular Formula | C19 H32 N2 O5 . C4 H11 N |
| Quantity | 250 mg |
| Synonym | Perindopril tert-butylamine, 2-Methylpropan-2-amine (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]propanoyl]octahydro-1H-indole-2-carboxylate, 1H-Indole-2-carboxylic acid','1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-','(2S,3aS,7aS)-','compd. with 2-methyl-2-propanamine (1:1), 1H-Indole-2-carboxylic acid','1-[2-[[1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-','[2S-[1[R*(R*)],2α,3aβ,7aβ]]-','compd. with 2-methyl-2-propanamine (1:1), 2-Propanamine','2-methyl-','(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate, 2-Propanamine','2-methyl-','[2S-[1[R*(R*)],2α,3aβ,7aβ]]-1-[2-[[1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate, Aceon, Butylaminiperindopril, Conversyl, Coversum, Coversyl, McN-A 2833-109, Perigard, Perindopril erbumine, Perindopril tert-butylamine salt, Perindopril-tert-butylamine, Perinodopril, Perinodpril erbimune, Prestarium, Procaptan, S 9490-3 |
| InChI Formula | InChI=1S/C19H32N2O5.C4H11N/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;1-4(2,3)5/h12-16,20H,4-11H2,1-3H3,(H,23,24);5H2,1-3H3/t12-,13-,14-,15-,16-;/m0./s1 |
| SMILES | CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)OCC.CC(C)(C)N |
| IUPAC Name | (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;2-methylpropan-2-amine |
| Molecular Weight (g/mol) | 441.6 |
| Formula Weight | 441.3203 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction