missing translation for 'onlineSavingsMsg'
Learn More

Perindopril t-Butylamine Salt, TRC

Product Code. 30415759
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 30415759

Brand: Toronto Research Chemicals TRCP28750550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perindopril tert-Butylamine
CAS 107133-36-8
Molecular Formula C19 H32 N2 O5 . C4 H11 N
Quantity 50 mg
Synonym Perindopril tert-butylamine, 2-Methylpropan-2-amine (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]propanoyl]octahydro-1H-indole-2-carboxylate, 1H-Indole-2-carboxylic acid','1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-','(2S,3aS,7aS)-','compd. with 2-methyl-2-propanamine (1:1), 1H-Indole-2-carboxylic acid','1-[2-[[1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-','[2S-[1[R*(R*)],2α,3aβ,7aβ]]-','compd. with 2-methyl-2-propanamine (1:1), 2-Propanamine','2-methyl-','(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate, 2-Propanamine','2-methyl-','[2S-[1[R*(R*)],2α,3aβ,7aβ]]-1-[2-[[1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate, Aceon, Butylaminiperindopril, Conversyl, Coversum, Coversyl, McN-A 2833-109, Perigard, Perindopril erbumine, Perindopril tert-butylamine salt, Perindopril-tert-butylamine, Perinodopril, Perinodpril erbimune, Prestarium, Procaptan, S 9490-3
InChI Formula InChI=1S/C19H32N2O5.C4H11N/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;1-4(2,3)5/h12-16,20H,4-11H2,1-3H3,(H,23,24);5H2,1-3H3/t12-,13-,14-,15-,16-;/m0./s1
SMILES CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)OCC.CC(C)(C)N
IUPAC Name (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;2-methylpropan-2-amine
Molecular Weight (g/mol) 441.6
Formula Weight 441.3203
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.