missing translation for 'onlineSavingsMsg'
Learn More

Perindopril-N-desethylpentanoate, TRC

Product Code. 30138098
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138098 1 g 1g
17982765 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138098 Supplier Toronto Research Chemicals Supplier No. TRCP2874901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685523-06-2
Synonym (2S,3aS,7aS)-1-[(2S)-2-aminopropanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Formula C12H20N2O3
Molecular Weight (g/mol) 240.3, 240.30
InChI Key BHUJIVBXVYHWJX-XKNYDFJKSA-N
SMILES [H][C@]12C[C@H](N(C(=O)[C@H](C)N)[C@@]1([H])CCCC2)C(O)=O
IUPAC Name (2S,3aS,7aS)-1-[(2S)-2-aminopropanoyl]-octahydro-1H-indole-2-carboxylic acid
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.