missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perindopril erbumine, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Perindopril erbumine (Perindopril tert-butylamine salt) is a potent ACE inhibitor of which is used to treat high blood pressure, heart failure or stable coronary artery disease.
Specifications
Specifications
| Chemical Name or Material | Perindopril erbumine |
| CAS | 107133-36-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H43N3O5 |
| Quantity | 100 mg |
| Synonym | Perindopril tert-butylamine saltS-9490 erbumine |
| Solubility Information | H2O : ≥ 50 mg/mL (113.22 mM) ∣DMSO : 10 mg/mL (22.64 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(C([C@@H](N[C@H](C(OCC)=O)CCC)C)=O)[C@@]2([H])CCCC[C@@]2([H])C1)O.CC(C)(C)N |
| Molecular Weight (g/mol) | 441.6 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction