missing translation for 'onlineSavingsMsg'
Learn More

Perindopril erbumine, MedChemExpress

Product Code. p-200101131
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16472036

Brand: Medchem Express HYB0130A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Perindopril erbumine (Perindopril tert-butylamine salt) is a potent ACE inhibitor of which is used to treat high blood pressure, heart failure or stable coronary artery disease.

Perindopril erbumine (Perindopril tert-butylamine salt) is a potent ACE inhibitor of which is used to treat high blood pressure, heart failure or stable coronary artery disease.

Specifications

Chemical Name or Material Perindopril erbumine
CAS 107133-36-8
Color White
Purity Grade Notes Research
Molecular Formula C23H43N3O5
Quantity 500 mg
Synonym Perindopril tert-butylamine saltS-9490 erbumine
Solubility Information H2O : ≥ 50 mg/mL (113.22 mM) ∣DMSO : 10 mg/mL (22.64 mM; Need ultrasonic)
SMILES O=C([C@H]1N(C([C@@H](N[C@H](C(OCC)=O)CCC)C)=O)[C@@]2([H])CCCC[C@@]2([H])C1)O.CC(C)(C)N
Molecular Weight (g/mol) 441.6
Formula Weight 441.6
Percent Purity 99.83%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.