missing translation for 'onlineSavingsMsg'
Learn More

Perilipin Antibody [DyLight 650], Novus Biologicals™

Product Code. 30730863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30730863 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730863 Supplier Novus Biologicals Supplier No. AF6615W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Perilipin Polyclonal antibody specifically detects Perilipin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein
Host Species Sheep
Immunogen E. coli-derived recombinant human Perilipin, Thr8-Ala145, Accession # O60240
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Lipid Droplets
Primary or Secondary Primary
Gene ID (Entrez) 5346
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.