missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-3/TIP47 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40765V
This item is not returnable.
View return policy
Description
Perilipin-3/TIP47 Polyclonal specifically detects Perilipin-3/TIP47 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Perilipin-3/TIP47 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1 | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Diabetes Research, Golgi Apparatus Markers, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 10226 | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PLIN3 | |
| A synthetic peptide made to a region within the N-terminus (within residues 1-100) of the mouse TIP47 protein. [Swiss-Prot# Q9DBG5] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction