missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-2/ADFP Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40877AF532
This item is not returnable.
View return policy
Description
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Perilipin-2/ADFP | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| Rabbit | |
| 0.1 mL | |
| 123 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PLIN2 | |
| A synthetic peptide made to a C-terminal region of mouse ADFP (within residues 350-425) [Swiss-ProtNo. P43883] | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction