missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1494), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Antigen | Perilipin-2/ADFP |
|---|---|
| Clone | ADFP/1494 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 2-4 ug/ml, Immunofluorescence 0.5-1.0 ug/ml |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243390
|
Novus Biologicals
NBP2-54597-20UG |
20 μg |
£211.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236800
|
Novus Biologicals
NBP2-54597-0.1MG |
0.1 mg |
£373.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Perilipin-2/ADFP | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 123 | |
| Recombinant human Adipophilin protein fragment (aa249-376) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 48 kDa |
| ADFP/1494 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 2-4 ug/ml, Immunofluorescence 0.5-1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| PLIN2 | |
| IgG2b κ | |
| Protein A or G purified | |
| Recognizes a protein of 48kDa, which is identified as Adipophilin. It belongs to the perilipin family, members of which coat intracellular lipid storage droplets. This protein is associated with the lipid globule surface membrane material, and maybe involved in development and maintenance of adipose tissue. However, it is not restricted to adipocytes as previously thought, but is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells, and tissues, such as lactating mammary gland, adrenal cortex, Sertoli and Leydig cells, and hepatocytes in alcoholic liver cirrhosis, suggesting that it may serve as a marker of lipid accumulation in diverse cell types and diseases. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title