missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Perilipin-2/ADFP Monoclonal antibody specifically detects Perilipin-2/ADFP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifications
Specifications
| Antigen | Perilipin-2/ADFP |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 788616 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human Perilipin-2, Met1-His437, Accession # Q99541 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?