missing translation for 'onlineSavingsMsg'
Learn More

Perilipin-2/ADFP Antibody (788616) [DyLight 755], Novus Biologicals™

Product Code. 30732362 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732362 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732362 Supplier Novus Biologicals Supplier No. FAB76341Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Perilipin-2/ADFP Monoclonal antibody specifically detects Perilipin-2/ADFP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin-2/ADFP
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 788616
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Host Species Mouse
Immunogen E. coli-derived recombinant human Perilipin-2, Met1-His437, Accession # Q99541
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Diabetes Research, Epigenetics, Hypoxia, Lipid and Metabolism, Lipid Droplets, metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 123
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.