missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin 1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518694
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine and rat based on sequence homology.
The protein encoded by this gene coats lipid storage droplets in adipocytes, thereby protecting them until they can be broken down by hormone-sensitive lipase. The encoded protein is the major cAMP-dependent protein kinase substrate in adipocytes and, when unphosphorylated, may play a role in the inhibition of lipolysis. Alternatively spliced transcript variants varying in the 5′ UTR, but encoding the same protein, have been found for this gene.
Specifications
| Perilipin 1 | |
| Polyclonal | |
| Unconjugated | |
| O60240, Q8CGN5 | |
| PLIN1 | |
| Synthetic peptide sequence (EPILGRAQYSQLRKK) corresponding to the C-terminus amino acids of PLIN | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PLIN1 | |
| Plin; perilipin; PERI; lipid droplet-associated protein | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 103968, 5346 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction