missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin 1 Polyclonal Antibody

Perilipin 1 Polyclonal Antibody
Brand: Invitrogen PA518693
This item is not returnable.
View return policy
Description
Perilipin 1 Polyclonal Antibody for Western Blot, IHC
Specifications
| Perilipin 1 | |
| Polyclonal | |
| Unconjugated | |
| Plin1 | |
| 6030432J05Rik; adipocyte lipid droplet binding protein; FPLD4; Lipid droplet-associated protein; PERI; PERIA; perilipin; perilipin 1; perilipin A; perilipin B; perilipin-1; PLIN; PLIN1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5346 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60240 | |
| Plin1 | |
| Synthetic peptide sequence (PREKPKRRVSDS) corresponding to the internal amino acids of PLIN (aa 488-500). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction