missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Perilipin 1 Monoclonal Antibody (GT2781)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527861
This item is not returnable.
View return policy
Description
Positive Control: mouse adipose Predicted Reactivity: Human (100%), Rat (92%), Pig (92%), Sheep (94%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Adipose tissue is an energy reserve in animals and is strictly regulated in nondomestic species. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), resistin, and perilipin. Perilipin is an intracellular neutral lipid droplet protein that is hormonally regulated. This protein is localized exclusively to the surface of lipid droplets. In response to lypotic stimuli, perilipin is phosphorylated by protein kinase A. Once activated, perilipin has inhibitory affects upon hormone-sensitive lipase (HSL), a protein that mediates the hydrolysis of triacylglycerol, the major form of stored energy in the body. Perilipin expression is limited to adipocytes and steroidogenic cells. There are currently two known isoforms, Perilipin A and B. Both of these proteins are encoded by a single-copy gene and are the result of differential splicing events.
Specifications
| Perilipin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative | |
| O60240, Q8CGN5 | |
| Plin1 | |
| Recombinant protein encompassing a sequence within the center region of human Perilipin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT2781 | |
| Unconjugated | |
| Plin1 | |
| 6030432J05Rik; adipocyte lipid droplet binding protein; FPLD4; Lipid droplet-associated protein; PERI; PERIA; perilipin; perilipin 1; perilipin A; perilipin B; perilipin-1; PLIN; PLIN1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 103968, 5346 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction