missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Perifosine

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | Perifosine |
| CAS | 157716-52-4 |
| Synonym | Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water and to 100mM in ethanol |
| Molecular Formula | C25H52NO4P |
| Formula Weight | 461.66 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction