missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Perifosine
GREENER_CHOICE

Product Code. 18723168
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18723168

Brand: Tocris Bioscience™ 6087/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PKB/Akt inhibitor; also modulates Kv2.1 current

Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perifosine
CAS 157716-52-4
Synonym Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in ethanol
Molecular Formula C25H52NO4P
Formula Weight 461.66
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.