missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Contec PeridoxRTU is a BPR and EPA authorised sporicide achieving greater than a log 3 reduction against bacterial and fungal spores in under 3 min
- A blend of hydrogen peroxide and peracetic acid, Contec PeridoxRTU is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts and spores
- PeridoxRTU's proprietary formulation contains surfactants for cleaning efficiency along with corrosion inhibitors for surface protection
- PeridoxRTU can be used to clean and disinfect surfaces without damage or loss of functionality
- Contec PeridoxRTU is filtered and filled in a controlled environment
- The sterile product is provided validated sterile to a Sterility Assurance Level of 10-6
- Provided double or single bagged, the product is designed for ease of entry into pharmaceutical cleanrooms
- Available as either a 0.9 L bottle with a push/pull cap for small areas or in 3.78 L capped containers for mop head saturation or when larger volumes are required
- Patented hyperactive chemistry makes it fast acting so save time spent of biodecontamination
- Validated sterile version available makes it suitable for Grade A and B cleanrooms
- Excellent material compatibility and will not damage common cleanroom materials
- Double/single bagged helps facilitate transfer disinfection into cleanroom
- Push/pull cap on 0.9 L eliminates droplets reducing the risk of inhalation and can be easily applied to a wipe or cleaning too
Specifications
Specifications
| Product Type | Sporicide |
| Form | Liquid |
| pH | Not Applicable |
| Sterility | Sterile |
| Appearance | Clear |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Composition | Hydrogen Peroxide and Peracetic Acid |
| Concentration | RTU |
| Disposable | Limited-use |
| Show More |
Safety and Handling
Product Identifier
- X6 Peridox Ready to Use Sporidical Disinfectant, 0 .9L, Double Bagged, Sterile
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P390-Absorb spillage to prevent material damage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Acetic acid, Hydrogen peroxide, Peracetic acid
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?