missing translation for 'onlineSavingsMsg'
Learn More

Peri-xanthenoxanthene, 95%, Thermo Scientific™

Product Code. 15284408 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15284408 1 g 1g
15294408 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284408 Supplier Thermo Scientific Acros Supplier No. 454890010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 191-28-6
Molecular Formula C20H10O2
Molecular Weight (g/mol) 282.3
InChI Key AMDQVKPUZIXQFC-UHFFFAOYSA-N
Synonym dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
PubChem CID 240798
SMILES C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 282.3
Percent Purity 95%
Chemical Name or Material Peri-xanthenoxanthene
compliance-icons
Product Identifier
  • Peri-xanthenoxanthene, 95%
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.