missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Performance Underbench ATEX Lab Freezer
GREENER_CHOICE

Product Code. 30464308 Shop All Liebherr Products
Change view
Click to view available options
Drawers:
3 transparent freezer drawers
Not included
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Drawers unitSize
30464308 Not included Each
30464305 3 transparent freezer drawers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464308 Supplier Liebherr™ Supplier No. SFTFG1501
  • ONLINE EXCLUSIVE PRICE
    £1890.67
    £1699.00 / Pack of 1
    Save £191.67
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Secure, accurate and ultra energy-efficient, spark-free interior, fridge storage, the Liebherr™ ATEX Underbench Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on.

  • Spark-Free underbench Lab Freezer for storage of solvents and reagents, width 600mm, gross capacity 101 L
  • Temperature range -9 / -30°C, +/- 5°C temperature stability tested to EN60068-3
  • White exterior, commercial-grade moulded polystyrol interior
  • static refrigeration with manual defrost
  • Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph, energy consumption 180 kWh/a
  • 73 Lnet capacity
  • 2 glass shelves
  • ATEX-certified spark-free interior
  • Multifunctional digital controller with high resolution display and connectivity options
  • Door lock, reversible hinges, antimicrobial lever handle
  • 40 kg shelf load

Specifications

Type ATEX freezer
Capacity (English) 26.7 gal.
Capacity (Metric) 101 L
Temperature Range -9°C to -30°C
Temperature Resolution Display 0.1 °C
Temperature Sensor NTC
Temperature Stability ±5 K
Door Style Reversible, Self-closing, Fitted Lock
No. of Doors 1
Door Material White Powder-coated steel
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Polystyrene
Interior Commercial-grade moulded polystyrol
Shelves 2
Drawers Not included
Certifications/Compliance ISO 9001, ISO 14001, CE, ATEX, DIN 13277
Style Underbench
Display Monochrome display in door
Color White
Condenser Type Low-maintenance rear-wall wire-on-tube
Control Type Touch control / digital display in door
For Use With (Application) Spark-free lab freezer storage
Ports 1 x 10 mm
Product Line Scientific Performance
Warranty 1 Year Parts and Labor
Defrost Manual
Model SFTfg 1501
Monitoring Options SmartCoolingHub
Connectivity LAN/WiFi module optional
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Frequency 50 Hz
Phase 1, single phase
Wattage 110 W
Energy Usage 180 kWh/a
Green Features Low energy consumption
Compressor CFC-free
Refrigerant R 600a
Shipping Weight (Metric) 38 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.