missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Performance Check Solution, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Bis(pentafluorophenyl)phenylphosphine | 5074-71-5 | 500μg/mL |
| Endrin | 72-20-8 | 500μg/mL |
| p,p'-DDT | 50-29-3 | 500μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Concentration | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation |
| Packaging | 2 mL Amber Prescored Glass Ampule |
| Quantity | 1 mL |
| Storage Requirements | Ambient |
| Type | Performance Check Solution |
Safety and Handling
Product Identifier
- Performance Check Solution
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dichloromethane
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction