Learn More
Performance Check Solution, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 5252-PCS
Code : 04
Additional Details : CAS Number : 75-09-2 Weight : 0.14000kg
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Bis(pentafluorophenyl)phenylphosphine | 5074-71-5 | 500μg/mL |
Endrin | 72-20-8 | 500μg/mL |
p,p'-DDT | 50-29-3 | 500μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
2 mL Amber Prescored Glass Ampule | |
Ambient |
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation | |
1 mL | |
Performance Check Solution |
Safety and Handling
- Performance Check Solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane
Product intended for laboratory use only