missing translation for 'onlineSavingsMsg'
Learn More

Perforin Mouse anti-Human, Non-human Primate, Clone: Pf-80/164, Novus Biologicals™

Product Code. 18346143
Change view
Click to view available options
Quantity:
2000 μg
500 μg
Unit Size:
2000µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346143 500 μg 500µg
18357033 2000 μg 2000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346143 Supplier Bio-Techne Supplier No. NBP318296500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Perforin Monoclonal antibody specifically detects Perforin in Human, Non-human Primate samples. It is validated for ELISA, ELISPOT
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perforin
Applications ELISA
Classification Monoclonal
Clone Pf-80/164
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Host Species Mouse
Immunogen Native human Perforin
Purification Method Protein G purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human, Non-human Primate
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.