missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Perforated Crate in Recycled HDPE

Perforated Crate 600 x 400 in recycled HDPE is ideal for storing all products requiring airflow and high durability in the intensive use cycle.

Brand:  Gilac™ G179705

Additional Details : Weight : 2.36000kg

 View more versions of this product

Product Code. 17958621

  • £28.37 / Each
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • 600 x 400 perforated crate in recycled HDPE suitable for food contact.
  • Stacking crate ideal for storing all products requiring airflow.
  • Highly durable in the intensive use cycle.
  • Ergonomic integrated handles for an easy and practical grip.
  • Compatible with 4 and 6-Wheel GILAC rolling dollies.
  • Recycled HDPE food crates offer the same level of professional quality as virgin HDPE crates.
  • Subject to very strict standards, recycled HDPE complies with all current food safety standards.
  • These new recycled HDPE crates also respond to changes in the anti-waste law.
  • By the end of 2022, purchasers from the State, local authorities and their groups will be obliged to acquire goods from reuse, or containing recycled materials.
Specifications

Specifications

15.9 gal.
Green
Rectangular
600 x 400 x 300 mm
23.6 in.
15.7 in.
11.8 in.
60 L
HDPE
Crate
600 mm
400 mm
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers