missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanoic acid, 95%, Thermo Scientific™

Product Code. 11382756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382756 25 g 25g
11372756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382756 Supplier Thermo Scientific Alfa Aesar Supplier No. L08862.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Perfluorooctanoic acid is used as a surfactant in the emulsion polymerization of fluoropolymers. It is used in fabrics and leather and in the production of floor waxes and waxed papers. The compound is also used in insulators for electric wires, planar etching of fused silica and fire fighting foam.

Solubility
Soluble in water. (3.4 g/L) at 20°C.

Notes
Store away from reducing agents and strong bases. Incompatible materials are Oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-67-1
Molecular Formula C8HF15O2
Molecular Weight (g/mol) 414.07
MDL Number MFCD00004174
InChI Key SNGREZUHAYWORS-UHFFFAOYSA-N
Synonym perfluorooctanoic acid, pentadecafluorooctanoic acid, perfluorocaprylic acid, pfoa, octanoic acid, pentadecafluoro, perfluoroctanoic acid, perfluoroheptanecarboxylic acid, pentadecafluoro-1-octanoic acid, pentadecafluoro-n-octanoic acid, perfluoro-n-octanoic acid
PubChem CID 9554
ChEBI CHEBI:35549
IUPAC Name pentadecafluorooctanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 58°C to 60°C
Density 1.7
Boiling Point 189°C to 192°C
Quantity 25 g
UN Number UN3261
Beilstein 1809678
Solubility Information Soluble in water. (3.4g/L) at 20°C.
Formula Weight 414.07
Percent Purity 95%
Chemical Name or Material Perfluorooctanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.