missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanoic Acid (50μg/mL in Methanol), TRC

Product Code. 30478099
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478099 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478099 Supplier Toronto Research Chemicals Supplier No. TRCKIT80351X1ML
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 335-67-1
Molecular Formula C8 H F15 O2
Quantity 1 mL
Synonym Octanoic acid, pentadecafluoro- (8CI,9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanoic acid, A 5717, EF 201, Eftop EF 201, NSC 95114, PFOA, Pentadecafluoro-n-octanoic acid, Pentadecafluorooctanoic acid, Perfluoro-1-heptanecarboxylic acid, Perfluorocaprylic acid, Perfluoroheptanecarboxylic acid, Perfluorooctanoic acid, n-Perfluorooctanoic acid, Perfluorooctanoic Acid, Octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-
InChI Formula InChI=1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoic acid
Molecular Weight (g/mol) 414.07
Formula Weight 413.9737
Concentration or Composition (by Analyte or Components) Perfluorooctanoic acid 50 μg/mL
Chemical Name or Material Perfluorooctanoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.